[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
T3445-1G
T3445-5G
76220-642EA
270.18
CAD
76220-642
76220-644
[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate
Formula:
C₁₇H₁₅F₆IO₃S Melting Pt: 183°C |
MDL Number:
MFCD20264880 CAS Number: 1204518-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...