2-(2-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC)

Supplier: TCI America

B5122-1G B5122-5G
76219-500EA 127.98 CAD
76219-500 76219-502
2-(2-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC)
2-(2-Biphenylyl)amino-9,9-dimethylfluorene
Formula: C₂₇H₂₃N
MDL Number: MFCD19440968
CAS Number: 1198395-24-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 118.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR