Methyl 3-Amino-2-fluorobenzoate ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Methyl 3-amino-2-fluorobenzoate, 3-Amino-2-fluorobenzoic acid methyl ester

M2868-1G M2868-200MG
TCM2868-1GEA 417.12 CAD
TCM2868-1G TCM2868-200MG
Methyl 3-Amino-2-fluorobenzoate ≥96.0% (by GC, titration analysis)
Methyl 3-Amino-2-fluorobenzoate
Formula: C₈H₈FNO₂
MDL Number: MFCD12923265
CAS Number: 1195768-18-3

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR