Tetrakis(4-hydroxyphenyl)ethylene ≥97.0% (by GC)

Supplier: TCI America

T3142-1G T3142-5G
TCT3142-1GEA 445.56 CAD
TCT3142-1G TCT3142-5G
Tetrakis(4-hydroxyphenyl)ethylene ≥97.0% (by GC)
Tetrakis(4-hydroxyphenyl)ethylene
Formula: C₂₆H₂₀O₄
Melting Pt: 323°C
MDL Number: MFCD28901376
CAS Number: 119301-59-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR