Lithium bis(nonafluorobutanesulfonyl)imide ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Bis(nonafluorobutanesulfonyl)imide Lithium salt, Bis(nonafluorobutanesulphonyl)imide Lithium salt

L0307-1G L0307-5G
TCL0307-1GEA 112.18 CAD
TCL0307-1G TCL0307-5G
Lithium bis(nonafluorobutanesulfonyl)imide ≥95.0% (by titrimetric analysis)
Lithium bis(nonafluorobutanesulfonyl)imide
Formula: C₈F₁₈LiNO₄S₂
MDL Number: MFCD06200837
CAS Number: 119229-99-1
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR