3-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5436-1G B5436-200MG
TCB5436-1GEA 353.92 CAD
TCB5436-1G TCB5436-200MG
3-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
3-Bromo-9,9-dimethyl-9H-fluorene
Formula: C₁₅H₁₃Br
MW: 273.17 g/mol
MDL Number: MFCD28955974
CAS Number: 1190360-23-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR