(4-Bromobutyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B5014-1G B5014-200MG
TCB5014-1GEA 984.34 CAD
TCB5014-1G TCB5014-200MG
(4-Bromobutyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(4-Bromobutyl)phosphonic acid
Formula: C₄H₁₀BrO₃P
MDL Number: MFCD00466589
CAS Number: 1190-14-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR