2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine dihydrochloride
I1057-1G
I1057-5G
TCI1057-1GEA
97.96
CAD
TCI1057-1G
TCI1057-5G
2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride ≥98.0% (by HPLC, titration analysis)
2-Isopropyl-4-(N-methylaminomethyl)thiazole dihydrochloride
Formula:
C₈H₁₄N₂S·₂HCl Storage Temperature: Refrigerator |
MDL Number:
MFCD11841218 CAS Number: 1185167-55-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...