Vinylsulfonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

V0134-1G V0134-5G
TCV0134-1GEA 176.96 CAD
TCV0134-1G TCV0134-5G
Vinylsulfonic acid ≥97.0% (by titrimetric analysis)
Vinylsulfonic acid
Formula: C₂H₄O₃S
Flash Pt: 159°C
Storage Temperature: Refrigerator
MDL Number: MFCD09743544
CAS Number: 1184-84-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (Bromination Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR