1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D3871-1G D3871-5G
TCD3871-1GEA 734.7 CAD
TCD3871-1G TCD3871-5G
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥98.0% (by HPLC)
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride
Formula: C₂₄H₆Br₂O₆
Melting Pt: 360°C
MDL Number: MFCD11870875
CAS Number: 118129-60-5

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR