4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone ≥98.0% (by GC)

Supplier: TCI America

P2372-1G P2372-5G
TCP2372-1GEA 60.04 CAD
TCP2372-1G TCP2372-5G
4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone ≥98.0% (by GC)
4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone
Formula: C₁₇H₃₀O
MDL Number: MFCD12911604
CAS Number: 117923-32-7

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR