9,9-Bis(4-methoxyphenyl)-9H-fluorene ≥98.0% (by GC)
Supplier: TCI America
B5617-1G
B5617-5G
TCB5617-1GEA
262.28
CAD
TCB5617-1G
TCB5617-5G
9,9-Bis(4-methoxyphenyl)-9H-fluorene ≥98.0% (by GC)
9,9-Bis(4-methoxyphenyl)-9H-fluorene
Formula:
C₂₇H₂₂O₂ |
CAS Number:
117766-40-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...