2,7-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5403-1G D5403-5G
76219-960EA 237 CAD
76219-960 76219-962
2,7-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)
2,7-Dibromo-9-heptyl-9H-carbazole
Formula: C₁₉H₂₁Br₂N
MDL Number: MFCD30742903
CAS Number: 1173071-58-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR