3,4,5-Tris(dodecyloxy)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America

T3624-1G T3624-200MG
TCT3624-1GEA 399.74 CAD
TCT3624-1G TCT3624-200MG
3,4,5-Tris(dodecyloxy)benzaldehyde ≥98.0% (by GC)
3,4,5-Tris(dodecyloxy)benzaldehyde
Formula: C₄₃H₇₈O₄
MDL Number: MFCD29917550
CAS Number: 117241-32-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR