3-Bromodibenzothiophene 5,5-Dioxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Bromodibenzosulpholane, 3-Bromodibenzo[b,d]thiophene 5,5-dioxide, 3-Bromodibenzothiophene sulfone, 3-Bromodibenzothiophene sulphone, 3-Bromodibenzosulfolane

B5268-1G B5268-200MG
TCB5268-1GEA 876.9 CAD
TCB5268-1G TCB5268-200MG
3-Bromodibenzothiophene 5,5-Dioxide ≥98.0% (by GC)
3-Bromodibenzothiophene 5,5-Dioxide
Formula: C₁₂H₇BrO₂S
MW: 295.15 g/mol
MDL Number: MFCD30742894
CAS Number: 116668-69-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 226.0 to 230.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR