4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
4,8-Bis((2-ethylhexyl)oxy)benzo[1,2-b:4,5-b']dithiophene
B4395-1G
B4395-200MG
B4395-5G
TCB4395-1GEA
295.46
CAD
TCB4395-1G
TCB4395-200MG
TCB4395-5G
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene
Formula:
C₂₆H₃₈O₂S₂ MW: 446.71 g/mol |
MDL Number:
MFCD17018551 CAS Number: 1160823-77-7 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (HPLC) | min. 95.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...