3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: N-(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)-[1,1'-biphenyl]-4-amine

B4887-1G B4887-5G
TCB4887-1GEA 140.46 CAD
TCB4887-1G TCB4887-5G
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole ≥98.0% (by HPLC, total nitrogen)
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole
Formula: C₃₆H₂₆N₂
MDL Number: MFCD22571697
CAS Number: 1160294-96-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 178.0 to 187.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR