N-(4-Bromophenyl)-4-biphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4753-1G B4753-5G
TCB4753-1GEA 58.46 CAD
TCB4753-1G TCB4753-5G
N-(4-Bromophenyl)-4-biphenylamine ≥98.0% (by GC)
N-(4-Bromophenyl)-4-biphenylamine
Formula: C₁₈H₁₄BrN
MDL Number: MFCD20486764
CAS Number: 1160294-93-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR