3-Bromo-6,9-diphenylcarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Bromo-6,9-diphenyl-9H-carbazole

B4795-1G B4795-5G
TCB4795-1GEA 59.57 CAD
TCB4795-1G TCB4795-5G
3-Bromo-6,9-diphenylcarbazole ≥98.0% (by GC)
3-Bromo-6,9-diphenylcarbazole
Formula: C₂₄H₁₆BrN
MDL Number: MFCD20257783
CAS Number: 1160294-85-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR