trans,trans-4-Propyl-4'-vinylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

P2317-25G P2317-5G
TCP2317-25GEA 797.9 CAD
TCP2317-25G TCP2317-5G
trans,trans-4-Propyl-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-Propyl-4'-vinylbicyclohexyl
Formula: C₁₇H₃₀
Storage Temperature: Refrigerator
MDL Number: MFCD11045059
CAS Number: 116020-44-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR