5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5057-1G B5057-5G
TCB5057-1GEA 90.06 CAD
TCB5057-1G TCB5057-5G
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene ≥98.0% (by GC)
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Formula: C₇H₂BrF₅O
MDL Number: MFCD03412225
CAS Number: 115467-07-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR