3-Hydroxy-1-methacryloyloxyadamantane ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
H1543-25G
H1543-5G
TCH1543-25GEA
707.52
CAD
TCH1543-25G
TCH1543-5G
3-Hydroxy-1-methacryloyloxyadamantane ≥98.0% (by GC)
3-Hydroxy-1-methacryloyloxyadamantane
Formula:
C₁₄H₂₀O₃ |
MDL Number:
MFCD13152044 CAS Number: 115372-36-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...