4,4,5,5-Tetramethyl-2-[10-(1-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane ≥97.0% (by GC)

Supplier: TCI America

T3306-1G T3306-5G
TCT3306-1GEA 316 CAD
TCT3306-1G TCT3306-5G
4,4,5,5-Tetramethyl-2-[10-(1-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-2-[10-(1-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane
Formula: C₃₀H₂₇BO₂
MDL Number: MFCD30470497
CAS Number: 1149804-35-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR