Pentafluorobenzyl methacrylate ≥98.0% (by GC) stabilised
Supplier: TCI America
|
Warning
|
Synonyms:
Methacrylic acid Pentafluorobenzyl ester
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
P2546-25G
P2546-5G
TCP2546-25GEA
1069.66
CAD
TCP2546-25G
TCP2546-5G
Pentafluorobenzyl methacrylate ≥98.0% (by GC) stabilised
Pentafluorobenzyl methacrylate
Formula:
C₁₁H₇F₅O₂ MW: 266.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00042329 CAS Number: 114859-23-3 |
Specification Test Results
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...