4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
9-Phenyl-10-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]anthracene, 4-(10-Phenylanthracen-9-yl)phenylboronic acid Pinacol ester
T3276-1G
T3276-200MG
TCT3276-1GEA
891.12
CAD
TCT3276-1G
TCT3276-200MG
4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane ≥95.0% (by HPLC, titration analysis)
4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane
Formula:
C₃₂H₂₉BO₂ MW: 456.39 g/mol |
CAS Number:
1143576-84-4 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...