9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)
Supplier: TCI America
B4666-1G
B4666-5G
TCB4666-1GEA
206.98
CAD
TCB4666-1G
TCB4666-5G
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene
Formula:
C₃₃H₃₀O₄ |
MDL Number:
MFCD22494984 CAS Number: 114205-89-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...