9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
P2376-1G
P2376-5G
TCP2376-1GEA
67.94
CAD
TCP2376-1G
TCP2376-5G
9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by HPLC, titration analysis)
9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formula:
C₂₄H₂₄BNO₂ |
MDL Number:
MFCD16621140 CAS Number: 1126522-69-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...