Epmedin C ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Baohuoside VI, 3-[[6-Deoxy-2-O-(6-deoxy-alpha-L-mannopyranosyl)-alpha-L-mannopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4-oxo-4H-chromen-7-yl beta-D-Glucopyranoside, 3-(((2S,3R,4R,5R,6S)-4,5-Dihydroxy-6-methyl-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

E1320-10MG E1320-50MG
76220-062EA 129.56 CAD
76220-062 76220-064
Epmedin C ≥95.0% (by HPLC)
Epmedin C
Formula: C₃₉H₅₀O₁₉
MW: 822.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039820
CAS Number: 110642-44-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Specific rotation [a]20/D -103 to -113 deg (C=0.2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR