N-Methoxy-N-methylbutyramide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
N-Methoxy-N-methylbutanamide
M3097-1G
M3097-5G
TCM3097-1GEA
67.94
CAD
TCM3097-1G
TCM3097-5G
N-Methoxy-N-methylbutyramide ≥98.0% (by GC)
N-Methoxy-N-methylbutyramide
Formula:
C₆H₁₃NO₂ MW: 131.18 g/mol |
MDL Number:
MFCD14707556 CAS Number: 109480-78-6 UN: 1993 ADR: 3 III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...