Biotin-LC-hydrazide ≥97.0% (by HPLC)

Supplier: TCI America

H1071-100MG H1071-25MG
TCH1071-100MGEA 396.58 CAD
TCH1071-100MG TCH1071-25MG
Biotin-LC-hydrazide ≥97.0% (by HPLC)
Biotin-LC-hydrazide
Formula: C₁₆H₂₉N₅O₃S
Storage Temperature: Freezer
MDL Number: MFCD00077659
CAS Number: 109276-34-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 18.00 to 19.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR