9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)

Supplier: TCI America

B4570-1G B4570-200MG
TCB4570-1GEA 1045.96 CAD
TCB4570-1G TCB4570-200MG
9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
9-Bromo-10-[4-(1-naphthyl)phenyl]anthracene
Formula: C₃₀H₁₉Br
MW: 459.39 g/mol
MDL Number: MFCD20486477
CAS Number: 1092390-01-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR