10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole ≥98.0% (by HPLC)

Supplier: TCI America

D5428-1G D5428-200MG
76219-972EA 571.8 CAD
76219-972 76219-974
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole ≥98.0% (by HPLC)
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole
Formula: C₂₄H₁₅N₃
Melting Pt: 350°C
MDL Number: MFCD21609440
CAS Number: 109005-10-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR