GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc ≥98.0% (by HPLC)

Supplier: TCI America

G0423-2MG G0423-5MG
TCG0423-2MGEA 461.36 CAD
TCG0423-2MG TCG0423-5MG
GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc ≥98.0% (by HPLC)
GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc
Formula: C₂₂H₃₈N₂O₁₅
Storage Temperature: Freezer
CAS Number: 108964-40-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR