2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4945-1G D4945-5G
76219-766EA 339.7 CAD
76219-766 76219-768
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide
Formula: C₃₀H₃₆Br₂N₂O₄
MDL Number: MFCD27923032
CAS Number: 1088205-02-0

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 243.0 to 247.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR