9-(3-Bromophenyl)-9H-fluoren-9-ol ≥98.0% (by GC)

Supplier: TCI America

B5615-1G B5615-5G
TCB5615-1GEA 278.08 CAD
TCB5615-1G TCB5615-5G
9-(3-Bromophenyl)-9H-fluoren-9-ol ≥98.0% (by GC)
9-(3-Bromophenyl)-9H-fluoren-9-ol
Formula: C₁₉H₁₃BrO
MDL Number: MFCD16659044
CAS Number: 1086641-47-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR