Triisopropyl[(trimethylsilyl)ethynyl]silane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3271-1ML T3271-5ML
TCT3271-1MLEA 109.02 CAD
TCT3271-1ML TCT3271-5ML
Triisopropyl[(trimethylsilyl)ethynyl]silane ≥97.0% (by GC)
Triisopropyl[(trimethylsilyl)ethynyl]silane
Formula: C₁₄H₃₀Si₂
MW: 254.56 g/mol
Boiling Pt: 57°C
MDL Number: MFCD28975120
CAS Number: 107474-02-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR