2-Phenoxyethyl methacrylate ≥85.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Ethylene Glycol Monophenyl ether methacrylate, Methacrylic acid 2-Phenoxyethyl ester

Stabilization: Stabilised with HQ + MEHQ

P2468-100G P2468-500G
TCP2468-100GEA 115.34 CAD
TCP2468-100G TCP2468-500G
2-Phenoxyethyl methacrylate ≥85.0% (by GC) stabilised
2-Phenoxyethyl methacrylate
Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Boiling Pt: 260°C
Flash Pt: 100°C
MDL Number: MFCD00053685
CAS Number: 10595-06-9
EINECS: 234-201-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR