9,9'-Bianthracene ≥98.0% (by HPLC)

Supplier: TCI America

B4095-1G B4095-5G
TCB4095-1GEA 398.16 CAD
TCB4095-1G TCB4095-5G
9,9'-Bianthracene ≥98.0% (by HPLC)
9,9'-Bianthracene
Formula: C₂₈H₁₈
Melting Pt: 306°C
MDL Number: MFCD00130216
CAS Number: 1055-23-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR