4,4',4''-Tris(diphenylamino)triphenylamine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N1,N1-Bis(4-(diphenylamino)phenyl)-N4,N4-diphenylbenzene-1,4-diamine, TDATA

T3062-1G T3062-200MG
TCT3062-1GEA 333.38 CAD
TCT3062-1G TCT3062-200MG
4,4',4''-Tris(diphenylamino)triphenylamine ≥97.0% (by HPLC, total nitrogen)
4,4',4''-Tris(diphenylamino)triphenylamine
Formula: C₅₄H₄₂N₄
MW: 746.96 g/mol
MDL Number: MFCD01310677
CAS Number: 105389-36-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 245.0 to 249.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR