2-Chloro-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

C3362-1G C3362-5G
TCC3362-1GEA 120.08 CAD
TCC3362-1G TCC3362-5G
2-Chloro-9H-carbazole ≥98.0% (by GC)
2-Chloro-9H-carbazole
Formula: C₁₂H₈ClN
MDL Number: MFCD01444890
CAS Number: 10537-08-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 243.0 to 247.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR