N-Carbobenzoxy-1,12-diaminododecane hydrochloride ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
C3472-1G
C3472-5G
TCC3472-1GEA
281.24
CAD
TCC3472-1G
TCC3472-5G
N-Carbobenzoxy-1,12-diaminododecane hydrochloride ≥98.0% (by HPLC)
N-Carbobenzoxy-1,12-diaminododecane hydrochloride
Formula:
C₂₀H₃₄N₂O₂·HCl |
MDL Number:
MFCD08272292 CAS Number: 1051420-16-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 173.0 to 178.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...