(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide ≥98.0% (by HPLC)

Supplier: TCI America

B5518-1G B5518-5G
76219-644EA 178.54 CAD
76219-644 76219-646
(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide ≥98.0% (by HPLC)
(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
Formula: C₂₅H₂₄N₂O₂
MDL Number: MFCD18910207
CAS Number: 105024-93-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 101.0 to 105.0 deg-C
Specific rotation [a]20/D +137 to +142 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR