N-(2-Propynyl)succinimide ≥98.0% (by GC)

Supplier: TCI America
Warning

P2191-1G P2191-200MG
TCP2191-1GEA 730.59 CAD
TCP2191-1G TCP2191-200MG
N-(2-Propynyl)succinimide ≥98.0% (by GC)
N-(2-Propynyl)succinimide
Formula: C₇H₇NO₂
Boiling Pt: 237°C
MDL Number: MFCD06740819
CAS Number: 10478-33-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR