Diethylene Glycol Monophenyl ether ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Phenyl Diglycol, 2-(2-Phenoxyethoxy)ethanol

D5139-25G D5139-500G
TCD5139-25GEA 46.59 CAD
TCD5139-25G TCD5139-500G
Diethylene Glycol Monophenyl ether ≥98.0% (by GC)
Diethylene Glycol Monophenyl ether
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 298°C
MDL Number: MFCD00045989
CAS Number: 104-68-7
EINECS: 203-227-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR