L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
LD-Heptose Peracetate
H1657-100MG
H1657-20MG
TCH1657-100MGEA
407.64
CAD
TCH1657-100MG
TCH1657-20MG
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate ≥98.0% (by GC)
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate
Formula:
C₁₉H₂₆O₁₃ MW: 462.40 g/mol Storage Temperature: Refrigerator |
CAS Number:
104486-80-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Specific rotation [a]20/D | +24.0 to +28.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...