Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate ≥98.0% (by GC)

Supplier: TCI America
Warning

E1149-100MG E1149-1G
TCE1149-100MGEA 96.38 CAD
TCE1149-100MG TCE1149-1G
Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate ≥98.0% (by GC)
Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate
Formula: C₁₃H₁₇ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00128924
CAS Number: 10443-70-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR