Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D4972-1G
D4972-5G
TCD4972-1GEA
448.72
CAD
TCD4972-1G
TCD4972-5G
Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione ≥98.0% (by GC, titration analysis)
Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione
Formula:
C₁₀H₄O₃S₂ Melting Pt: 263°C |
CAS Number:
1043023-52-4 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...