1,2-Bis(5-methyl-2-benzoxazolyl)ethylene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1,2-Bis(5-methylbenzo[d]oxazol-2-yl)ethene

B4250-25G B4250-5G
TCB4250-25GEA 798.37 CAD
TCB4250-25G TCB4250-5G
1,2-Bis(5-methyl-2-benzoxazolyl)ethylene ≥98.0% (by HPLC, total nitrogen)
1,2-Bis(5-methyl-2-benzoxazolyl)ethylene
Formula: C₁₈H₁₄N₂O₂
MW: 290.32 g/mol
MDL Number: MFCD00408425
CAS Number: 1041-00-5
EINECS: 213-866-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR