Atipamezole hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2956-100MG A2956-25MG
76218-764EA 943.26 CAD
76218-764 76218-766
Atipamezole hydrochloride ≥98.0% (by HPLC, titration analysis)
Atipamezole hydrochloride
Formula: C₁₄H₁₆N₂·HCl
MDL Number: MFCD06407819
CAS Number: 104075-48-1
Merck Index: 14,00861

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR