5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T3485-1G T3485-5G
TCT3485-1GEA 99.54 CAD
TCT3485-1G TCT3485-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde ≥98.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
Formula: C₁₁H₁₅BO₃S
MDL Number: MFCD09266186
CAS Number: 1040281-83-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR